(tert-Butyldimethylsilyl)acetylen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

399264-1G 399264-5G
SIAL399264-1GEA 84.2 EUR
SIAL399264-1G SIAL399264-5G
(tert-Butyldimethylsilyl)acetylen, Sigma-Aldrich®
(tert-Butyldimethylsilyl)acetylen
Formel: C₈H₁₆Si
Molecular Weight: 140,3 g/mol
MDL: MFCD00191877
CAS-Nummer: 86318-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR