3,9-Diethyl-3,6,9-triazaundecan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1-(2-(Diethylamino)ethyl)-N2,N2-diethylethane-1,2-diamine

424072-25ML 424072-5ML
SIAL424072-25MLEA 334 EUR
SIAL424072-25ML SIAL424072-5ML
3,9-Diethyl-3,6,9-triazaundecan, Sigma-Aldrich®
3,9-Diethyl-3,6,9-triazaundecan
Formel: C₁₂H₂₉N₃
Molecular Weight: 215,38 g/mol
MDL: MFCD00039880
CAS-Nummer: 123-12-6
EINECS: 204-603-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR