Diphenyliodoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

43088-25G 43088-5G
SIAL43088-25GEA 244 EUR
SIAL43088-25G SIAL43088-5G
Diphenyliodoniumchlorid, Sigma-Aldrich®
Diphenyliodonium chloride
Formel: C₁₂H₁₀ClI
Molecular Weight: 316,57 g/mol
MDL: MFCD00011909
CAS-Nummer: 1483-72-3
EINECS: 216-049-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR