2,6-Dichlorisonicotinsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,6-Dichloropyridine-4-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
456543-1G
456543-5G
SIAL456543-1GEA
25.1
EUR
SIAL456543-1G
SIAL456543-5G
2,6-Dichlorisonicotinsäure, Sigma-Aldrich®
2,6-Dichlorisonicotinsäure
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol |
MDL:
MFCD00234147 CAS-Nummer: 5398-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...