6-Brompicolinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

484652-1G 484652-5G
SIAL484652-1GEA 54.7 EUR
SIAL484652-1G SIAL484652-5G
6-Brompicolinsäure, Sigma-Aldrich®
6-Brompicolinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202,01 g/mol
MDL: MFCD00093197
CAS-Nummer: 21190-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR