(S)-(-)-Hydroxypyrrolidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine

56437-1ML 56437-5ML
SIAL56437-1MLEA 93.1 EUR
SIAL56437-1ML SIAL56437-5ML
(S)-(-)-Hydroxypyrrolidin, Sigma-Aldrich®
(S)-(-)-Hydroxypyrrolidin
Formel: C₄H₉NO
Molecular Weight: 87,12 g/mol
MDL: MFCD00192426
CAS-Nummer: 100243-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR