Indium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Indiumtrichlorid

Unter Argon in Ampullen verpackt

44349.06 44349.22
44349.06EA 122 EUR
44349.06 44349.22
Indium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Indium(III)chlorid

Purity: 99.999% CAS No.: 10025-82-8 Molecular Formula: Cl3In Formula Weight: 221.18 Physical Form: Powder, Ampouled under argon Density: 3.460 Appearance: White Odour: Odourless Melting Point: 586° Boiling Point: 800° Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Formel: Cl₃In
Molecular Weight: 221,18 g/mol
Siedepunkt: 800 °C
Schmelzpunkt: 586 °C
Dichte: 3,460
Lagertemperatur: Raumtemperatur
MDL: MFCD00011058
CAS-Nummer: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 14,4958

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR