3-Fluorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluoropyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

638757-10G 638757-1G
SIAL638757-10GEA 132 EUR
SIAL638757-10G SIAL638757-1G
3-Fluorisonicotinsäure, Sigma-Aldrich®
3-Fluorisonicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD03789251
CAS-Nummer: 393-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR