5-Brom-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine

M-Clarity™ Qualitätsstufe = MQ100

641359-1G 641359-5G
SIAL641359-1GEA 26 EUR
SIAL641359-1G SIAL641359-5G
5-Brom-2-pyridincarbonitril, Sigma-Aldrich®
5-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
MDL: MFCD00234144
CAS-Nummer: 97483-77-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR