Methylendiphosphonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Medronic acid
64255-1G-F
64255-25G-F
64255-5G-F
SIAL64255-1G-FEA
212
EUR
SIAL64255-1G-F
SIAL64255-25G-F
SIAL64255-5G-F
Methylendiphosphonsäure, Sigma-Aldrich®
Methylenediphosphonic acid
Formel:
CH₆O₆P₂ Molecular Weight: 176 g/mol |
MDL:
MFCD00002139 CAS-Nummer: 1984-15-2 EINECS: 217-851-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...