2-Methoxy-4-nitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

658324-1G 658324-5G
SIAL658324-1GEA 31.4 EUR
SIAL658324-1G SIAL658324-5G
2-Methoxy-4-nitrobenzaldehyd, Sigma-Aldrich®
2-Methoxy-4-nitrobenzaldehyde
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD02683052
CAS-Nummer: 136507-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR