2,3,5-Trifluorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,5-Trifluoropyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

660647-1G 660647-250MG
SIAL660647-1GEA 131 EUR
SIAL660647-1G SIAL660647-250MG
2,3,5-Trifluorisonicotinsäure, Sigma-Aldrich®
2,3,5-Trifluorisonicotinsäure
Formel: C₆H₂F₃NO₂
Molecular Weight: 177,08 g/mol
MDL: MFCD03840205
CAS-Nummer: 675602-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR