8-Anilino-1-naphthalinsulfonsäure, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid
M-Clarity™ Qualitätsstufe = MQ200
A1028-100G
A1028-25G
A1028-5G
A1028-100GEA
823
EUR
A1028-100G
A1028-25G
A1028-5G
8-Anilino-1-naphthalinsulfonsäure, Sigma-Aldrich®
8-Anilino-1-naphthalinsulfonsäure
Formel:
C₁₆H₁₃NO₃S Schmelzpunkt: 215…217 °C (Zers., Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00003998 CAS-Nummer: 82-76-8 EINECS: 201-438-7 |
|
Spezifikation Testergebnisse
Appearance (Colour) |
Conforms to Requirements Gray to dark green or black |
Appearance (Form) | Powder |
Solubility (Colour) | Dark Green to Dark Brown to Black |
Solubility (Turbidity) 50 mg/ml, DMF |
Clear to Turbid |
Proton NMR spectrum | Conforms to Structure |
Solvent Content | ≤0.5% |
Purity (HPLC) | ≥97% |
Recommended Retest Period | 3 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...