Aminoacetaldehyddiethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal

M-Clarity™ Qualitätsstufe = MQ100

A37200-100ML A37200-25ML
SIALA37200-100MLEA 288 EUR
SIALA37200-100ML SIALA37200-25ML
Aminoacetaldehyddiethylacetal, Sigma-Aldrich®
Aminoacetaldehyddiethylacetal
Formel: C₆H₁₅NO₂
Molecular Weight: 133,19 g/mol
MDL: MFCD00008136
CAS-Nummer: 645-36-3
EINECS: 211-439-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR