3-Sulfolen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-sulpholene, Butadiene sulphone

M-Clarity™ Qualitätsstufe = MQ100

B84505-100G B84505-500G
SIALB84505-100GEA 42.8 EUR
SIALB84505-100G SIALB84505-500G
3-Sulfolen, Sigma-Aldrich®
3-Sulfolen
Formel: C₄H₆O₂S
Molecular Weight: 118,16 g/mol
MDL: MFCD00005481
CAS-Nummer: 77-79-2
EINECS: 201-059-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR