Cystaminiumdichlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cystaminium dichloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

C8707-100G C8707-25G
SIALC8707-100GEA 1450 EUR
SIALC8707-100G SIALC8707-25G
Cystaminiumdichlorid, Sigma-Aldrich®
Cystaminiumdichlorid
Formel: C₄H₁₂N₂S₂·2HCl
Molecular Weight: 225.20 g/mol
MDL: MFCD00012905
CAS-Nummer: 56-17-7
EINECS: 200-260-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR