1,4-Diacryloylpiperazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1'-(Piperazine-1,4-diyl)bis(prop-2-en-1-one)

D1538-10G D1538-5G
SIALD1538-10GEA 612 EUR
SIALD1538-10G SIALD1538-5G
1,4-Diacryloylpiperazin, Sigma-Aldrich®
1,4-Diacryloylpiperazin
Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194.23 g/mol
MDL: MFCD00077661
CAS-Nummer: 6342-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR