Dibenzylphosphonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dibenzyl phosphite
M-Clarity™ Qualitätsstufe = MQ100
D36607-100G
D36607-25G
D36607-5G
SIALD36607-100GEA
237
EUR
SIALD36607-100G
SIALD36607-25G
SIALD36607-5G
Dibenzylphosphonat, Sigma-Aldrich®
Dibenzylphosphonat
Formel:
C₁₄H₁₅O₃P Molecular Weight: 262,25 g/mol |
MDL:
MFCD00004774 CAS-Nummer: 17176-77-1 EINECS: 241-226-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...