N,N-Dimethylglycin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N,N-Dimethylglycine HCl
M-Clarity™ Qualitätsstufe = MQ200
D6382-25G
D6382-5G
SIALD6382-25GEA
213
EUR
SIALD6382-25G
SIALD6382-5G
N,N-Dimethylglycin Hydrochlorid, Sigma-Aldrich®
N,N-Dimethylglycin Hydrochlorid
Formel:
C₄H₁₀ClNO₂ Molecular Weight: 139,58 g/mol |
MDL:
MFCD00012610 CAS-Nummer: 2491-06-7 EINECS: 219-648-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...