(±)-Ethyl-2-brombutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-bromobutyrate, Ethyl 2-bromobutanoate

M-Clarity™ Qualitätsstufe = MQ100

E13814-100G E13814-5G
SIALE13814-100GEA 63.6 EUR
SIALE13814-100G SIALE13814-5G
(±)-Ethyl-2-brombutyrat, Sigma-Aldrich®
(±)-Ethyl-2-brombutyrat
Formel: C₆H₁₁BrO₂
Molecular Weight: 195,06 g/mol
MDL: MFCD00000155
CAS-Nummer: 533-68-6
EINECS: 208-574-6
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR