N-Methylformanilid, Sigma-Aldrich®

Lieferant: Merck

M46802-100G M46802-500G
SIALM46802-100GEA 57 EUR
SIALM46802-100G SIALM46802-500G
N-Methylformanilid, Sigma-Aldrich®
N-Methylformanilid
Formel: C₈H₉NO
Molecular Weight: 135.17 g/mol
MDL: MFCD00003283
CAS-Nummer: 93-61-8
EINECS: 202-262-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR