2-Mercapto-2-thiazolin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
M-Clarity™ Qualitätsstufe = MQ200
M6204-100G
M6204-500G
SIALM6204-100GEA
50.4
EUR
SIALM6204-100G
SIALM6204-500G
2-Mercapto-2-thiazolin, Sigma-Aldrich®
2-Mercapto-2-thiazolin
Formel:
C₃H₅NS₂ Molecular Weight: 119,21 g/mol |
MDL:
MFCD00126013 CAS-Nummer: 96-53-7 EINECS: 202-512-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...