N-Methylphenethylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

M68423-100ML M68423-25ML M68423-5ML
SIALM68423-100MLEA 491 EUR
SIALM68423-100ML SIALM68423-25ML SIALM68423-5ML
N-Methylphenethylamin, Sigma-Aldrich®
N-Methylphenethylamin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
MDL: MFCD00008291
CAS-Nummer: 589-08-2
EINECS: 209-632-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR