Methoxyacetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

M9009-100G-A M9009-25G-A M9009-5G-A
SIALM9009-100G-AEA 588 EUR
SIALM9009-100G-A SIALM9009-25G-A SIALM9009-5G-A
Methoxyacetonitril, Sigma-Aldrich®
Methoxyacetonitril
Formel: C₃H₅NO
Molecular Weight: 71,08 g/mol
MDL: MFCD00001892
CAS-Nummer: 1738-36-9
EINECS: 217-092-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR