Methylendiphosphonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Medronic acid
M9508-250MG
M9508-10G
M9508-1G
M9508-5G
SIALM9508-250MGEA
63.9
EUR
SIALM9508-250MG
SIALM9508-10G
SIALM9508-1G
SIALM9508-5G
Methylendiphosphonsäure, Sigma-Aldrich®
Methylenediphosphonic acid
Formel:
CH₆O₆P₂ Molecular Weight: 176 g/mol |
MDL:
MFCD00002139 CAS-Nummer: 1984-15-2 EINECS: 217-851-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...