Pyridin-2-aldoxim methochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-2-carboxaldoxime methochloride, Pralidoxime Chloride, 2-PAM chloride, 2-((Hydroxyimino)methyl)-1-methylpyridin-1-ium chloride

P9053-100G P9053-25G P9053-5G
SIALP9053-100GEA 1250 EUR
SIALP9053-100G SIALP9053-25G SIALP9053-5G
Pyridin-2-aldoxim methochlorid, Sigma-Aldrich®
Pyridin-2-aldoxim methochlorid
Formel: C₇H₉ClN₂O
Molecular Weight: 172,61 g/mol
MDL: MFCD00011981
CAS-Nummer: 51-15-0
EINECS: 200-080-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR