Bis(trimethylsilyl)acetylen
Lieferant: CHEMPUR SILANE
SILS02400.10
SILS02400.50
SILS02400.250
SILNSILS02400.10EA
0
EUR
SILNSILS02400.10
SILNSILS02400.50
SILNSILS02400.250
Bis(trimethylsilyl)acetylen
Bis(trimethylsilyl)acetylen
Formel:
C₈H₁₈Si₂ |
MDL:
MFCD00008276 CAS-Nummer: 14630-40-1 EINECS: 238-671-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...