(tert-Butyldimethylsilyl)acetylen

Lieferant: CHEMPUR SILANE

SILS03339.10 SILS03339.25
SILNSILS03339.10EA 0 EUR
SILNSILS03339.10 SILNSILS03339.25
(tert-Butyldimethylsilyl)acetylen
(tert-Butyldimethylsilyl)acetylen
Formel: C₈H₁₆Si
Molecular Weight: 140,3 g/mol
MDL: MFCD00191877
CAS-Nummer: 86318-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR