1-Phenyl-2-(trimethylsilyl)acetylen

Lieferant: CHEMPUR SILANE

Synonyme: Trimethyl(phenylethynyl)silane

SILS13785.10 SILS13785.25
SILNSILS13785.10EA 0 EUR
SILNSILS13785.10 SILNSILS13785.25
1-Phenyl-2-(trimethylsilyl)acetylen
1-Phenyl-2-(trimethylsilyl)acetylen
Formel: C₁₁H₁₄Si
Molecular Weight: 174.32 g/mol
MDL: MFCD00054860
CAS-Nummer: 2170-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR