(Triisopropylsilyl)acetylen

Lieferant: CHEMPUR SILANE

Synonyme: Ethynyltriisopropylsilane

SILS18000.5 SILS18000.25
SILNSILS18000.5EA 0 EUR
SILNSILS18000.5 SILNSILS18000.25
(Triisopropylsilyl)acetylen
(Triisopropylsilyl)acetylen
Formel: C₁₁H₂₂Si
Molecular Weight: 182,38 g/mol
MDL: MFCD00075452
CAS-Nummer: 89343-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR