Thallium(I)iodid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Thalliumiodid
Unter Argon in Ampullen verpackt
35722.09
35722.18
35722.30
35722.09EA
100
EUR
35722.09
35722.18
35722.30
Thallium(I)iodid, ultra trocken ≥99,999% (Metall-Basis)
Thallium(I)iodid
Purity: 99.999%
CAS No.: 7790-30-9
Molecular Formula: TlI
Formula Weight: 331.27
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 7.1
Appearance: Yellow
Odour: Odourless
Melting Point: 440°
Boiling Point: 824°
Merck Reference: 14,9264
UN No.: 1707
MDL No.: MFCD00011279
Formel:
ITl Molecular Weight: 331,29 g/mol Siedepunkt: 824 °C Schmelzpunkt: 440 °C Dichte: 7,1 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011279 CAS-Nummer: 7790-30-9 EINECS: 232-199-7 UN: 1707 ADR: 6.1,II Merck Index: 14,9264 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...