di-Bortrioxid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Borsäureanhydrid
A11707.36
A11707.0E
A11707.36EA
50.5
EUR
A11707.36
A11707.0E
di-Bortrioxid ≥99%
di-Bortrioxid
Purity: 99%
CAS No.: 1303-86-2
Molecular Formula: B2O3
Formula Weight: 69.62
Odour: Odourless
Melting Point: ca 450°
Boiling Point: 1860°
Merck Reference: 14,1337
MDL No.: MFCD00011315
Formel:
B₂O₃ Molecular Weight: 69,62 g/mol Siedepunkt: 1860 °C Schmelzpunkt: ∼450 °C Dichte: 2,46 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011315 CAS-Nummer: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 14,1337 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...