Tetrachlorsilan ≥99,998% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Siliciumtetrachlorid, Silicium(IV)chlorid
40148.AC
40148.AE
40148.ACEA
56.2
EUR
40148.AC
40148.AE
Tetrachlorsilan ≥99,998% (Metall-Basis)
Tetrachlorsilan
Purity: 99.998%
CAS No.: 10026-04-7
Molecular Formula: Cl4Si
Formula Weight: 169.90
Physical Form: Liquid
Density: 1.483
Appearance: Colourless
Odour: Acrid
Melting Point: -70°
Boiling Point: 57.6°
Merck Reference: 14,8500
UN No.: 1818
MDL No.: MFCD00011229
Formel:
Cl₄Si Molecular Weight: 169,9 g/mol Siedepunkt: 57,6 °C Schmelzpunkt: –70 °C Dichte: 1,483 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011229 CAS-Nummer: 10026-04-7 EINECS: 233-054-0 UN: 1818 ADR: 8,II Merck Index: 14,8500 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...