Kieselgel, Porengröße: 60 Å 0.060-0.2 mm (70-230 mesh)
Lieferant: Thermo Fisher Scientific
Achtung
L14002.0B L14002.0E
L14002.0BEA 99 EUR
L14002.0B L14002.0E
Kieselgel, Porengröße: 60 Å 0.060-0.2 mm (70-230 mesh)
Silica gel
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and removing impurities. It is used as stationary base in column chromatography.
CAS No.: 7631-86-9 Odour: Odourless Merck Reference: 14,8493 MDL No.: MFCD00011232
Formel: SiO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
Merck Index: 14,8493