Blei(II)tellurid 99,99 Metals basis
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
36317.09
36317.18
36317.09EA
110
EUR
36317.09
36317.18
Blei(II)tellurid 99,99 Metals basis
Blei(II)tellurid
Purity: 99.99%
CAS No.: 1314-91-6
Molecular Formula: PbTe
Formula Weight: 334.79
Physical Form: -325 Mesh Powder
Density: 8.164
Melting Point: 917°
Merck Reference: 14,5422
UN No.: 3284
MDL No.: MFCD00016274
In semiconductor research, photoconductor cells
Formel:
PbTe Molecular Weight: 334.8 g/mol Schmelzpunkt: 917 °C Dichte: 8.164 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016274 CAS-Nummer: 1314-91-6 EINECS: 215-247-1 UN: 3284 ADR: 6.1 III Merck Index: 14,5422 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...