Zinn(II)tellurid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

89186.09 89186.18
89186.09EA 117 EUR
89186.09 89186.18
Zinn(II)tellurid ≥99,999% (Metall-Basis)
Zinn(II)tellurid

Purity: 99.999% CAS No.: 12040-02-7 Molecular Formula: SnTe Formula Weight: 246.29 Physical Form: 3-12mm Pieces Density: 6.48 Melting Point: 780° UN No.: 3288 MDL No.: MFCD00049543

Formel: SnTe
Molecular Weight: 246.31 g/mol
Schmelzpunkt: 780 °C
Dichte: 6.48
Lagertemperatur: Raumtemperatur
MDL: MFCD00049543
CAS-Nummer: 12040-02-7
EINECS: 234-914-8
UN: 3288
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR