Antimon(V)chlorid ≥99,997% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Antimonpentachlorid
17570.14
17570.22
17570.14EA
148
EUR
17570.14
17570.22
Antimon(V)chlorid ≥99,997% (Metall-Basis)
Antimon(V)chlorid
Purity: 99.997%
CAS No.: 7647-18-9
Molecular Formula: Cl5Sb
Formula Weight: 299.02
Physical Form: Liquid
Density: 2.336
Refractive Index: 1.601
Sensitivity: Moisture Sensitive
Appearance: Red-yellow
Odour: Pungent
Melting Point: 2.8°
Boiling Point: 79°/22mm
Merck Reference: 14,695
UN No.: 1730
MDL No.: MFCD00011213
Formel:
Cl₅Sb Molecular Weight: 299,02 g/mol Siedepunkt: 79 °C (22 mmHg) Schmelzpunkt: 2,8 °C Dichte: 2,336 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011213 CAS-Nummer: 7647-18-9 EINECS: 231-601-8 UN: 1730 ADR: 8,II Merck Index: 14,695 |