Phosphor(V)chlorid ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
11849.14 11849.22
11849.14EA 147 EUR
11849.14 11849.22
Phosphor(V)chlorid ≥99,99% (Metall-Basis)
Phosphor(V)chlorid
Purity: 99.998% CAS No.: 10026-13-8 Molecular Formula: PCl5 Formula Weight: 208.24 Physical Form: Solid Density: 2.114 Appearance: Yellow Odour: Pungent Melting Point: 162° subl. Merck Reference: 14,7351 UN No.: 1806 MDL No.: MFCD00011439
Formel: Cl₅P
Molecular Weight: 208,24 g/mol
Schmelzpunkt: 162 °C (Subl.)
Dichte: 2,114
Lagertemperatur: Raumtemperatur
MDL: MFCD00011439
CAS-Nummer: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8,II
Merck Index: 14,7351