Phosphorylchlorid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Phosphor(V)oxychlorid, Phosphoroxidtrichlorid
10525.30
10525.A1
10525.30EA
57.8
EUR
10525.30
10525.A1
Phosphorylchlorid ≥99%
Phosphorylchlorid
Purity: 99%
CAS No.: 10025-87-3
Molecular Formula: Cl3OP
Formula Weight: 153.33
Physical Form: Liquid
Density: 1.645
Refractive Index: 1.460
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pungent
Melting Point: 1.25°
Boiling Point: 105.8°
Merck Reference: 14,7349
UN No.: 1810
MDL No.: MFCD00011443
Formel:
POCl₃ Molecular Weight: 153,33 g/mol Siedepunkt: 105,8 °C Schmelzpunkt: 1,25 °C Dichte: 1,645 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011443 CAS-Nummer: 10025-87-3 EINECS: 233-046-7 UN: 1810 ADR: 6.1,I Merck Index: 14,7349 |