Bismut(III)tellurid ≥99,98% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dibismuth tritelluride
44077.14
44077.22
44077.14EA
306
EUR
44077.14
44077.22
Bismut(III)tellurid ≥99,98% (Metall-Basis)
Bismut(III)tellurid
Purity: 99.98%
CAS No.: 1304-82-1
Molecular Formula: Bi2Te3
Formula Weight: 800.76
Physical Form: Powder
Appearance: Grey
Odour: Odourless
Melting Point: 573°
UN No.: 3284
MDL No.: MFCD00014201
Formel:
Bi₂Te₃ Molecular Weight: 800,76 g/mol Schmelzpunkt: 573 °C Dichte: 7,69 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014201 CAS-Nummer: 1304-82-1 EINECS: 215-135-2 UN: 3284 ADR: 6.1,III Merck Index: 12,01331 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...