Tellurtetraiodid ≥99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Tellurium(IV) iodide
87795.06
87795.14
87795.06EA
71.3
EUR
87795.06
87795.14
Tellurtetraiodid ≥99% (Metall-Basis)
Tellurtetraiodid
Purity: 99%
CAS No.: 7790-48-9
Molecular Formula: TeI4
Formula Weight: 635.22
Physical Form: Crystalline
Density: 5.403
Melting Point: 280°
Boiling Point: dec.
Solubility: Decomposes slowly in cold water to TeO2 and HI
Merck Reference: 14,9127
UN No.: 2923
MDL No.: MFCD00049578
Formel:
I₄Te Molecular Weight: 635,22 g/mol Siedepunkt: Zersetzung Schmelzpunkt: 280 °C Dichte: 5,403 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049578 CAS-Nummer: 7790-48-9 EINECS: 232-210-5 UN: 3260 ADR: 8,II Merck Index: 14,9127 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...