Tellurdioxid, tech.

Lieferant: Thermo Scientific
Achtung

10608.18 10608.30
10608.18EA 132 EUR
10608.18 10608.30
Tellurdioxid, tech.
Tellurdioxid

CAS No.: 7446-07-3 Molecular Formula: TeO2 Formula Weight: 159.60 Physical Form: Powder Density: 5.8 Odour: Odourless Merck Reference: 14,9123 MDL No.: MFCD00011263

Formel: O₂Te
Molecular Weight: 159,6 g/mol
Siedepunkt: 1245 °C (1013 hPa)
Schmelzpunkt: 733 °C
Dichte: 5,8
Lagertemperatur: Raumtemperatur
MDL: MFCD00011263
CAS-Nummer: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,9123

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR