Tellurdioxid ≥99,995% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
11821.09
11821.18
11821.30
11821.09EA
85.4
EUR
11821.09
11821.18
11821.30
Tellurdioxid ≥99,995% (Metall-Basis)
Tellurdioxid
Purity: 99.995%
CAS No.: 7446-07-3
Molecular Formula: TeO2
Formula Weight: 159.60
Physical Form: Powder
Density: 5.8
Melting Point: 733°
Merck Reference: 14,9123
MDL No.: MFCD00011263
Formel:
O₂Te Siedepunkt: 1245 °C (1013 hPa) Schmelzpunkt: 733 °C Dichte: 5,8 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011263 CAS-Nummer: 7446-07-3 EINECS: 231-193-1 Merck Index: 14,9123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...