Tellurdioxid ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
87813.14
87813.22
87813.14EA
96.8
EUR
87813.14
87813.22
Tellurdioxid ≥99,99% (Metall-Basis)
Tellurdioxid
Purity: 99.99%
CAS No.: 7446-07-3
Molecular Formula: TeO2
Formula Weight: 159.60
Physical Form: Powder
Density: 5.8
Melting Point: 733°
Merck Reference: 14,9123
MDL No.: MFCD00011263
Formel:
O₂Te Molecular Weight: 159,6 g/mol Siedepunkt: 1245 °C (1013 hPa) Schmelzpunkt: 733 °C Dichte: 5,8 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011263 CAS-Nummer: 7446-07-3 EINECS: 231-193-1 Merck Index: 14,9123 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...