2-Brom-1-indanol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-hydroxyindane
A13511.18
A13511.09
A13511.18EA
107
EUR
A13511.18
A13511.09
2-Brom-1-indanol ≥99%
2-Brom-1-indanol
Purity: 99%
CAS No.: 5400-80-6
Molecular Formula: C9H9BrO
Formula Weight: 213.08
Melting Point: 128-132°
Beilstein Reference: 2046877
Formel:
C₉H₉BrO Molecular Weight: 213.07 g/mol Siedepunkt: 290 Schmelzpunkt: 128…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003798 CAS-Nummer: 5400-80-6 EINECS: 226-442-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...