2-Brom-1-indanol
Lieferant: FLUOROCHEM
Synonyme:
2-Bromo-1-hydroxyindane
003061-100G
003061-25G
FLUO003061-100GEA
0
EUR
FLUO003061-100G
FLUO003061-25G
2-Brom-1-indanol
2-Brom-1-indanol
Formel:
C₉H₉BrO Molecular Weight: 213.07 g/mol Siedepunkt: 290 Schmelzpunkt: 131…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003798 CAS-Nummer: 5400-80-6 EINECS: 226-442-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...