2-Indanon 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one
164110100
164110500
ACRO164110100EA
78.4
EUR
ACRO164110100
ACRO164110500
2-Indanon 97%
2-Indanon
Formel:
C₉H₈O Molecular Weight: 132.16 g/mol Siedepunkt: 220…225 °C (1013 hPa) Schmelzpunkt: 50…57 °C Flammpunkt: 100 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00003792 CAS-Nummer: 615-13-4 EINECS: 210-410-3 |
Spezifikation Testergebnisse
Appearance | Light yellow to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 50 to 57 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...