5-Methoxy-1-indanon ≥98%

Lieferant: Thermo Fisher Scientific

L11165.03 L11165.06
L11165.03EA 33.1 EUR
L11165.03 L11165.06
5-Methoxy-1-indanon ≥98%
5-Methoxy-1-indanon

Purity: 98% CAS No.: 5111-70-6 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 108-110° Beilstein Reference: 1282137

Formel: C₁₀H₁₀O₂
Molecular Weight: 162.19 g/mol
Siedepunkt: 145…150 °C (0.2 mmHg)
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003789
CAS-Nummer: 5111-70-6
EINECS: 225-838-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR