1,3-Indandion 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,3-Dioxoindan
122090050
122090250
122091000
ACRO122090050EA
33.9
EUR
ACRO122090050
ACRO122090250
ACRO122091000
1,3-Indandion 97%
1,3-Indandion
Formel:
C₉H₆O₂ Molecular Weight: 146.15 g/mol Siedepunkt: 283 Schmelzpunkt: 129…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003779 CAS-Nummer: 606-23-5 EINECS: 210-109-7 |
Spezifikation Testergebnisse
Appearance | Light yellow to beige to golden Powder |
Infrared spectrum | Conforms |
Melting point | 124 to 131 °C |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...