5-Aminoindan ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Aminoindane

L02504.09 L02504.18
L02504.09EA 73.8 EUR
L02504.09 L02504.18
5-Aminoindan ≥98%
5-Aminoindan

Purity: 98+% CAS No.: 24425-40-9 Molecular Formula: C9H11N Formula Weight: 133.19 Flash Point: 110°(230°F) Melting Point: 32-37° Boiling Point: 248-250° Beilstein Reference: 2082278 UN No.: 2811 MDL No.: MFCD00003803

Formel: C₉H₁₁N
Molecular Weight: 133,19 g/mol
Siedepunkt: 248…250 °C
Schmelzpunkt: 32…37 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003803
CAS-Nummer: 24425-40-9
EINECS: 246-241-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR